Trimethylpropylammonium bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N,N,N-Trimethylpropan-1-aminium bis((trifluoromethyl)sulphonyl)amide, N,N,N-Trimethylpropan-1-aminium bis((trifluoromethyl)sulfonyl)amide

T2761-25G T2761-5G
TCT2761-25GEA 739.44 CAD
TCT2761-25G TCT2761-5G
Trimethylpropylammonium bis(trifluoromethanesulfonyl)imide ≥98.0% (by titrimetric analysis)
Trimethylpropylammonium bis(trifluoromethanesulfonyl)imide

CAS Number: 268536-05-6
MDL Number: MFCD08458920
Molecular Formula: C8H16F6N2O4S2
Molecular Weight: 382.34
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Melting point (°C): 19

Formula: C₈H₁₆F₆N₂O₄S₂
MW: 382.34 g/mol
Melting Pt: 19 °C
MDL Number: MFCD08458920
CAS Number: 268536-05-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8cbbb468310d3134d8997ce04fdd4d9b8c91aa02&vid=8cbbb468310d3134d8997ce04fdd4d9b8c91aa02&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR