2',4',5'-Trifluoroacetophenone ≥97.0% (by GC)
Supplier: TCI America
T2758-1G
T2758-5G
TCT2758-1GEA
44.56
CAD
TCT2758-1G
TCT2758-5G
2',4',5'-Trifluoroacetophenone ≥97.0% (by GC)
2',4',5'-Trifluoroacetophenone
CAS Number: 129322-83-4
MDL Number: MFCD00061193
Molecular Formula: C8H5F3O
Molecular Weight: 174.12
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Very Pale Yellow
Flash Point (°C): 71
Specific Gravity (20/20): 1.35
Formula:
C₈H₅F₃O Density: 1.35 (20 °C) Flash Pt: 71 °C |
MDL Number:
MFCD00061193 CAS Number: 129322-83-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...