1H-Pyrazole-4-boronic acid pinacol ester ≥97.0% (by GC)

Supplier: TCI America

T2756-1G T2756-5G
TCT2756-1GEA 62.88 CAD
TCT2756-1G TCT2756-5G
1H-Pyrazole-4-boronic acid pinacol ester ≥97.0% (by GC)
1H-Pyrazole-4-boronic acid pinacol ester

CAS Number: 269410-08-4
MDL Number: MFCD03453063
Molecular Formula: C9H15BN2O2
Molecular Weight: 194.04
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 147


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-(4-Pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Pyrazoleboronic Acid Pinacol Ester

Formula: C₉H₁₅BN₂O₂
Melting Pt: 147 °C
MDL Number: MFCD03453063
CAS Number: 269410-08-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a98e7728419bc8c1364fe26a50d9ee230ff308c4&vid=a98e7728419bc8c1364fe26a50d9ee230ff308c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR