Tetramethylammonium fluoride tetrahydrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T2754-25G T2754-5G
TCT2754-25GEA 161.16 CAD
TCT2754-25G TCT2754-5G
Tetramethylammonium fluoride tetrahydrate ≥98.0% (by titrimetric analysis)
Tetramethylammonium fluoride tetrahydrate

CAS Number: 17787-40-5
MDL Number: MFCD00149968
Molecular Formula: C4H12FN
Molecular Weight: 93.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 38
Flash Point (°C): 113
Storage Temperature: 0-10°C

Formula: C₄H₁₂FN·4H₂O
Melting Pt: 38 °C
Flash Pt: 113 °C
MDL Number: MFCD00149968
CAS Number: 17787-40-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1cc1043d0319517a332a63c48bc5e4f1f740448&vid=b1cc1043d0319517a332a63c48bc5e4f1f740448&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR