Trimorpholinophosphine oxide ≥98.0%

Supplier: TCI America

Synonyms: Phosphoric Trimorpholide

T2748-25G T2748-5G
TCT2748-25GEA 176.96 CAD
TCT2748-25G TCT2748-5G
Trimorpholinophosphine oxide ≥98.0%
Trimorpholinophosphine oxide

CAS Number: 4441-12-7
MDL Number: MFCD00047406
Molecular Formula: C12H24N3O4P
Molecular Weight: 305.31
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 191


Other Product Names / Synonyms: Phosphoric Trimorpholide

Formula: C₁₂H₂₄N₃O₄P
MW: 305.31 g/mol
Melting Pt: 191 °C
MDL Number: MFCD00047406
CAS Number: 4441-12-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=498c876349b81a675c72b4f8e6f9cbda7192d188&vid=498c876349b81a675c72b4f8e6f9cbda7192d188&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR