2,3,5,6-Tetrafluoro-1,4-benzenedimethanol ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 2,3,5,6-Tetrafluoro-1,4-bis(hydroxymethyl)benzene, 2,3,5,6-Tetrafluoro-p-xylene-alpha,alpha'-diol, 2,3,5,6-Tetrafluoro-alpha,alpha'-dihydroxy-p-xylene

T2732-25G T2732-5G
TCT2732-25GEA 404.95 CAD
TCT2732-25G TCT2732-5G
2,3,5,6-Tetrafluoro-1,4-benzenedimethanol ≥97.0% (by GC)
2,3,5,6-Tetrafluoro-1,4-benzenedimethanol

CAS Number: 92339-07-6
MDL Number: MFCD00229147
Molecular Formula: C8H6F4O2
Molecular Weight: 210.13
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 129


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Monomers
Other Product Names / Synonyms: 2,3,5,6-Tetrafluoro-1,4-bis(hydroxymethyl)benzene, 2,3,5,6-Tetrafluoro-alpha,alpha'-dihydroxy-p-xylene, 2,3,5,6-Tetrafluoro-p-xylene-alpha,alpha'-diol

Formula: C₈H₆F₄O₂
MW: 210.13 g/mol
Melting Pt: 129 °C
MDL Number: MFCD00229147
CAS Number: 92339-07-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b70f9e1407333c37dd39bd2663cb7552623ffb23&vid=b70f9e1407333c37dd39bd2663cb7552623ffb23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR