Thieno[2,3-b]thiophene ≥97.0% (by GC)

Supplier: TCI America

T2729-1G T2729-5G
TCT2729-1GEA 172.22 CAD
TCT2729-1G TCT2729-5G
Thieno[2,3-b]thiophene ≥97.0% (by GC)
Thieno[2,3-b]thiophene

CAS Number: 250-84-0
MDL Number: MFCD02824126
Molecular Formula: C6H4S2
Molecular Weight: 140.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 226
Melting point (°C): 7
Flash Point (°C): 93
Specific Gravity (20/20): 1.34


Product Categories: Materials Chemistry, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 1,6-Dithiapentalene

Formula: C₆H₄S₂
Boiling Pt: 226 °C
Melting Pt: 7 °C
Density: 1.34 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD02824126
CAS Number: 250-84-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=253bbe5fb08fefe03c2661529429c3af92b25a5f&vid=253bbe5fb08fefe03c2661529429c3af92b25a5f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR