1,3,5-Tris(4-aminophenyl)benzene ≥93.0% (by HPLC, titration analysis)

Supplier: TCI America

T2728-5G T2728-25G
TCT2728-5GEA 566.76 CAD
TCT2728-5G 76220-526
1,3,5-Tris(4-aminophenyl)benzene ≥93.0% (by HPLC, titration analysis)
1,3,5-Tris(4-aminophenyl)benzene

CAS Number: 118727-34-7
Molecular Formula: C24H21N3
Molecular Weight: 351.45
Purity/Analysis Method: >93.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 165

Formula: C₂₄H₂₁N₃
MW: 351.45 g/mol
Melting Pt: 165 °C
MDL Number: MFCD18207723
CAS Number: 118727-34-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d13a9a625d1b2d501d6fcf5e1307f98b61e2205&vid=7d13a9a625d1b2d501d6fcf5e1307f98b61e2205&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR