1-(2,3,4-Trimethoxybenzyl)piperazine Dihydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
T2726-25G
T2726-5G
TCT2726-25GEA
191.18
CAD
TCT2726-25G
TCT2726-5G
1-(2,3,4-Trimethoxybenzyl)piperazine Dihydrochloride ≥98.0% (by HPLC, total nitrogen)
1-(2,3,4-Trimethoxybenzyl)piperazine Dihydrochloride
CAS Number: 13171-25-0
MDL Number: MFCD00243086
Molecular Formula: C14H22N2O3
Molecular Weight: 339.26
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 228
Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: Trimetazidine Dihydrochloride
Formula:
C₁₄H₂₂N₂O₃ Melting Pt: 228 °C |
MDL Number:
MFCD00243086 CAS Number: 13171-25-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...