2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone ≥98.0%

Supplier: TCI America

Synonyms: 1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanone

T2722-25G T2722-5G
TCT2722-25GEA 281.24 CAD
TCT2722-25G TCT2722-5G
2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone ≥98.0%
2',4'-Difluoro-2-(1H-1,2,4-triazol-1-yl)acetophenone

CAS Number: 86404-63-9
MDL Number: MFCD02093825
Molecular Formula: C10H7F2N3O
Molecular Weight: 223.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 109


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(2,4-Difluorobenzoylmethyl)-1H-1,2,4-triazole

Formula: C₁₀H₇F₂N₃O
MW: 223.18 g/mol
Melting Pt: 109 °C
MDL Number: MFCD02093825
CAS Number: 86404-63-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c92e05f46d70ccce255669a4880c78e44ff6a54&vid=0c92e05f46d70ccce255669a4880c78e44ff6a54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR