Baicalein ≥98.0%
Supplier: TCI America
Synonyms:
5,6,7-Trihydroxyflavone, 5,6,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 5,6,7-Trihydroxy-2-phenylchromen-4-one
CAS Number: 491-67-8
MDL Number: MFCD00017459
Molecular Formula: C15H10O5
Molecular Weight: 270.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 265
Storage Temperature: 0-10°C
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 5,6,7-Trihydroxyflavone
Formula:
C₁₅H₁₀O₅ Melting Pt: 265 °C |
MDL Number:
MFCD00017459 CAS Number: 491-67-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...