3,4,5-Trichloropyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T2714-25G T2714-5G
TCT2714-25GEA 183.28 CAD
TCT2714-25G TCT2714-5G
3,4,5-Trichloropyridine ≥98.0% (by GC, titration analysis)
3,4,5-Trichloropyridine

CAS Number: 33216-52-3
MDL Number: MFCD00051685
Molecular Formula: C5H2Cl3N
Molecular Weight: 182.43
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 215
Melting point (°C): 73

Formula: C₅H₂Cl₃N
Boiling Pt: 215 °C
Melting Pt: 73 °C
MDL Number: MFCD00051685
CAS Number: 33216-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82b462ab5d0cd6ba5a79db9bede902922f1e1f5f&vid=82b462ab5d0cd6ba5a79db9bede902922f1e1f5f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR