Tangeretin ≥95.0% (by HPLC)
Supplier: TCI America
T2708-100MG
T2708-10MG
TCT2708-100MGEA
178.54
CAD
TCT2708-100MG
TCT2708-10MG
Tangeretin ≥95.0% (by HPLC)
Tangeretin
CAS Number: 481-53-8
MDL Number: MFCD00017438
Molecular Formula: C20H20O7
Molecular Weight: 372.37
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 155
Storage Temperature: 0-10°C
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Aromatic Polyketides, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: 4',5,6,7,8-Pentamethoxyflavone
Formula:
C₂₀H₂₀O₇ Melting Pt: 155 °C |
MDL Number:
MFCD00017438 CAS Number: 481-53-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...