Trimesic acid trimethyl ester ≥98.0%

Supplier: TCI America

Synonyms: Trimethyl 1,3,5-benzenetricarboxylate, Trimethyl benzene-1,3,5-tricarboxylate, 1,3,5-Benzenetricarboxylic acid trimethyl ester

T2707-25G T2707-5G
TCT2707-25GEA 204.09 CAD
TCT2707-25G TCT2707-5G
Trimesic acid trimethyl ester ≥98.0%
Trimesic acid trimethyl ester

CAS Number: 2672-58-4
MDL Number: MFCD00008434
Molecular Formula: C12H12O6
Molecular Weight: 252.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 230
Melting point (°C): 146
Lambda max.: 282 nm (CH3CN)


Other Product Names / Synonyms: Trimesic Acid Trimethyl Ester, Trimethyl Trimesate, 1,3,5-Benzenetricarboxylic Acid Trimethyl Ester

Formula: C₁₂H₁₂O₆
MW: 252.22
Boiling Pt: 230 °C
Melting Pt: 146 °C
MDL Number: MFCD00008434
CAS Number: 2672-58-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89fc7b50dd7d16085743fe120ee8c955c2f7cca6&vid=89fc7b50dd7d16085743fe120ee8c955c2f7cca6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR