Teprenone [mixture of (5E,9E,13E)- and (5Z,9E,13E)- isomers] ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
6,10,14,18-Tetramethyl-5,9,13,17-nonadecatetraen-2-one, Geranylgeranylacetone
CAS Number: 6809-52-5
MDL Number: MFCD00869547
Molecular Formula: C23H38O
Molecular Weight: 330.56
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 160
Specific Gravity (20/20): 0.89
Storage Temperature: <0°C
Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: Geranylgeranylacetone, 6,10,14,18-Tetramethyl-5,9,13,17-nonadecatetraen-2-one
Formula:
C₂₃H₃₈O MW: 330.56 g/mol Boiling Pt: 160 °C Density: 0.89 (20 °C) |
MDL Number:
MFCD00869547 CAS Number: 6809-52-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...