2,4,6-Tri(9H-carbazol-9-yl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
Supplier: TCI America
T2700-1G
T2700-5G
TCT2700-1GEA
364.98
CAD
TCT2700-1G
TCT2700-5G
2,4,6-Tri(9H-carbazol-9-yl)-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
2,4,6-Tri(9H-carbazol-9-yl)-1,3,5-triazine
CAS Number: 134984-37-5
Molecular Formula: C39H24N6
Molecular Weight: 576.66
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Color: White
Formula:
C₃₉H₂₄N₆ MW: 576.66 g/mol |
MDL Number:
MFCD18207722 CAS Number: 134984-37-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...