3,3',5,5'-Tetrabromo-2,2'-bithiophene ≥98.0% (by GC)

Supplier: TCI America

T2699-1G T2699-25G T2699-5G
TCT2699-1GEA 61.62 CAD
TCT2699-1G TCT2699-25G TCT2699-5G
3,3',5,5'-Tetrabromo-2,2'-bithiophene ≥98.0% (by GC)
3,3',5,5'-Tetrabromo-2,2'-bithiophene

CAS Number: 125143-53-5
Molecular Formula: C8H2Br4S2
Molecular Weight: 481.84
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 140

Formula: C₈H₂Br₄S₂
Melting Pt: 140 °C
MDL Number: MFCD00114807
CAS Number: 125143-53-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eaa18c55e96c27d1a775d1ab9308374748677d32&vid=eaa18c55e96c27d1a775d1ab9308374748677d32&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR