Tricyclo[6.4.0.0(2,7)]dodecane-1,8:2,7-tetracarboxylic dianhydride ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Octahydrobiphenylene-4a,8b:4b,8a-tetracarboxylic Dianhydride, Octahydro-4a,8b:4b,8a-bis(methanooxymethano)biphenylene-9,11,12,14-tetraone
CAS Number: 738-90-9
Molecular Formula: C16H16O6
Molecular Weight: 304.30
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 283
Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: Octahydrobiphenylene-4a,8b:4b,8a-tetracarboxylic Dianhydride, Octahydro-4a,8b:4b,8a-bis(methanooxymethano)biphenylene-9,11,12,14-tetraone
Formula:
C₁₆H₁₆O₆ MW: 304.30 g/mol Melting Pt: 283 °C |
CAS Number:
738-90-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...