1,2,3,4-Tetramethyl-1,2,3,4-cyclobutanetetracarboxylic dianhydride
Supplier: TCI America
T2697-1G
T2697-5G
TCT2697-1GEA
159.58
CAD
TCT2697-1G
TCT2697-5G
1,2,3,4-Tetramethyl-1,2,3,4-cyclobutanetetracarboxylic dianhydride
1,2,3,4-Tetramethyl-1,2,3,4-cyclobutanetetracarboxylic dianhydride
CAS Number: 64198-16-9
Molecular Formula: C12H12O6
Molecular Weight: 252.22
Form: Crystal
Color: White
Melting point (°C): 380
Formula:
C₁₂H₁₂O₆ Melting Pt: 380 °C |
CAS Number:
64198-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...