Tetrabutylammonium acetate ≥90.0%

Supplier: TCI America

Synonyms: Tetra-n-butylammonium acetate

T2694-25G T2694-100G
TCT2694-25GEA 190.07 CAD
TCT2694-25G TCT2694-100G
Tetrabutylammonium acetate ≥90.0%
Tetrabutylammonium acetate

CAS Number: 10534-59-5
MDL Number: MFCD00043208
Molecular Formula: C18H39NO2
Molecular Weight: 301.52
Purity/Analysis Method: >90.0% (T)
Form: Paste
Color: White
Melting point (°C): 98

Formula: C₁₈H₃₉NO₂
Melting Pt: 98 °C
MDL Number: MFCD00043208
CAS Number: 10534-59-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=405882971c2595b7b46a8ce7bf9cbe65b34f36dc&vid=405882971c2595b7b46a8ce7bf9cbe65b34f36dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR