2-[4-(Trifluoromethyl)phenyl]pyridine ≥98.0%

Supplier: TCI America

Synonyms: 2-(4-(Trifluoromethyl)phenyl)pyridine

T2688-1G T2688-5G
TCT2688-1GEA 175.38 CAD
TCT2688-1G TCT2688-5G
2-[4-(Trifluoromethyl)phenyl]pyridine ≥98.0%
2-[4-(Trifluoromethyl)phenyl]pyridine

CAS Number: 203065-88-7
MDL Number: MFCD08235168
Molecular Formula: C12H8F3N
Molecular Weight: 223.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 77

Formula: C₁₂H₈F₃N
MW: 223.20 g/mol
Melting Pt: 77 °C
MDL Number: MFCD08235168
CAS Number: 203065-88-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13f3ea0ddc0dc2c4fffc8d0199a40f1477dbc469&vid=13f3ea0ddc0dc2c4fffc8d0199a40f1477dbc469&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR