4,5,6,7-Tetrahydrothieno[3,2-c]pyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T2686-25G T2686-5G
TCT2686-25GEA 304.94 CAD
TCT2686-25G TCT2686-5G
4,5,6,7-Tetrahydrothieno[3,2-c]pyridine ≥98.0% (by GC, titration analysis)
4,5,6,7-Tetrahydrothieno[3,2-c]pyridine

CAS Number: 54903-50-3
MDL Number: MFCD03839944
Molecular Formula: C7H9NS
Molecular Weight: 139.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 75
Specific Gravity (20/20): 1.18
Storage Temperature: 0-10°C

Formula: C₇H₉NS
Boiling Pt: 75 °C
Density: 1.18 (20 °C)
MDL Number: MFCD03839944
CAS Number: 54903-50-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eda892a5c9b0ac7d2b85d2df12352722d39ba56c&vid=eda892a5c9b0ac7d2b85d2df12352722d39ba56c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR