(3-Glycidoxypropyl)triethoxysilane ≥97.0% (by GC)

Supplier: TCI America

T2675-25G T2675-5G
TCT2675-25GEA 73.93 CAD
TCT2675-25G TCT2675-5G
(3-Glycidoxypropyl)triethoxysilane ≥97.0% (by GC)
(3-Glycidoxypropyl)triethoxysilane

CAS Number: 2602-34-8
MDL Number: MFCD01311705
Molecular Formula: C12H26O5Si
Molecular Weight: 278.42
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 135
Flash Point (°C): 118
Specific Gravity (20/20): 1.01


Product Categories: Materials Chemistry, Silane Coupling Agents, Monomers
Other Product Names / Synonyms: Glycidyl 3-Triethoxysilylpropyl Ether

Formula: C₁₂H₂₆O₅Si
Boiling Pt: 135 °C
Density: 1.01 (20 °C)
Flash Pt: 118 °C
MDL Number: MFCD01311705
CAS Number: 2602-34-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0f9d2b17c0dd9c805e75996f9a8f045e8d962e3&vid=f0f9d2b17c0dd9c805e75996f9a8f045e8d962e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR