5,6,7,8-Tetrahydronaphthalene-1-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T2672-25G T2672-5G
TCT2672-25GEA 443.03 CAD
TCT2672-25G TCT2672-5G
5,6,7,8-Tetrahydronaphthalene-1-carboxylic acid ≥98.0% (by GC, titration analysis)
5,6,7,8-Tetrahydronaphthalene-1-carboxylic acid

CAS Number: 4242-18-6
MDL Number: MFCD01464030
Molecular Formula: C11H12O2
Molecular Weight: 176.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 152

Formula: C₁₁H₁₂O₂
Melting Pt: 152 °C
MDL Number: MFCD01464030
CAS Number: 4242-18-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=68be53c6f95ed7f5821369ae60ff60530e87b33a&vid=68be53c6f95ed7f5821369ae60ff60530e87b33a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR