4-(Trimethylsilyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

T2664-1G T2664-5G
TCT2664-1GEA 109.02 CAD
TCT2664-1G TCT2664-5G
4-(Trimethylsilyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(Trimethylsilyl)phenylboronic acid

CAS Number: 17865-11-1
MDL Number: MFCD00093348
Molecular Formula: C9H15BO2Si
Molecular Weight: 194.11
Form: Crystal
Color: White


Other Product Names / Synonyms: 4-(Trimethylsilyl)benzeneboronic Acid

Formula: C₉H₁₅BO₂Si
MDL Number: MFCD00093348
CAS Number: 17865-11-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=655fb4d2f0fa825880c6f79ccdbde89f204211f6&vid=655fb4d2f0fa825880c6f79ccdbde89f204211f6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR