3-(Trimethylsilyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

T2663-5G T2663-1G
TCT2663-5GEA 309.68 CAD
TCT2663-5G TCT2663-1G
3-(Trimethylsilyl)phenylboronic acid (contains varying amounts of Anhydride)
3-(Trimethylsilyl)phenylboronic acid

CAS Number: 177171-16-3
MDL Number: MFCD03093884
Molecular Formula: C9H15BO2Si
Molecular Weight: 194.11
Form: Crystal
Color: White


Other Product Names / Synonyms: 3-(Trimethylsilyl)benzeneboronic Acid

Formula: C₉H₁₅BO₂Si
MDL Number: MFCD03093884
CAS Number: 177171-16-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72bb2e84155aadc76914d3d5c5d526906888a9e1&vid=72bb2e84155aadc76914d3d5c5d526906888a9e1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR