2,4,6-Triisopropylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
2,4,6-Triisopropylbenzeneboronic acid, (2,4,6-Triisopropylphenyl)boronic acid
T2654-1G
T2654-5G
TCT2654-1GEA
94.17
CAD
TCT2654-1G
TCT2654-5G
2,4,6-Triisopropylphenylboronic acid (contains varying amounts of Anhydride)
2,4,6-Triisopropylphenylboronic acid
CAS Number: 154549-38-9
MDL Number: MFCD02683099
Molecular Formula: C15H25BO2
Molecular Weight: 248.17
Form: Crystal
Color: White
Melting point (°C): 165
Other Product Names / Synonyms: 2,4,6-Triisopropylbenzeneboronic Acid
Formula:
C₁₅H₂₅BO₂ MW: 248.16 g/mol Melting Pt: 165 °C |
MDL Number:
MFCD02683099 CAS Number: 154549-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...