D-ɑ-Tocopherol succinate ≥97.0%

Supplier: TCI America

Synonyms: Vitamin E Succinate, D-α-Tocopherol Succinate, 4-Oxo-4-(((R)-2,5,7,8-tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-yl)oxy)butanoic acid

T2628-25G T2628-5G
TCT2628-25GEA 200.91 CAD
TCT2628-25G TCT2628-5G
D-ɑ-Tocopherol succinate ≥97.0%
D-alpha-Tocopherol succinate

CAS Number: 4345-03-3
MDL Number: MFCD00072055
Molecular Formula: C33H54O5
Molecular Weight: 530.79
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 75
Specific rotation [a]20/D: 4 deg (C=8, EtOH)
Lambda max.: 286 nm (EtOH)Storage Temperature: 0-10°C


Product Categories: Bioscience, Vitamin Derivatives, Terpenes (Others)
Other Product Names / Synonyms: Vitamin E Succinate

Formula: C₃₃H₅₄O₅
MW: 530.79 g/mol
Melting Pt: 75 °C
MDL Number: MFCD00072055
CAS Number: 4345-03-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1af645309a1b630fb75c94342bd2b43054a0ce44&vid=1af645309a1b630fb75c94342bd2b43054a0ce44&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR