Trimethylolpropane tris(3-mercaptopropionate) ≥85.0% (by GC)

Supplier: TCI America

Synonyms: 1,1,1-Tris(3-mercaptopropionyloxymethyl)propane

T2619-25G T2619-500G
TCT2619-25GEA 44.56 CAD
TCT2619-25G TCT2619-500G
Trimethylolpropane tris(3-mercaptopropionate) ≥85.0% (by GC)
Trimethylolpropane tris(3-mercaptopropionate)

CAS Number: 33007-83-9
MDL Number: MFCD00046842
Molecular Formula: C15H26O6S3
Molecular Weight: 398.55
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 220
Flash Point (°C): 96
Specific Gravity (20/20): 1.22


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,1,1-Tris(3-mercaptopropionyloxymethyl)propane

Formula: C₁₅H₂₆O₆S₃
MW: 398.55 g/mol
Boiling Pt: 220 °C
Density: 1.22 (20 °C)
Flash Pt: 96 °C
MDL Number: MFCD00046842
CAS Number: 33007-83-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a15687812137d17cc1e5037082238499bee33c0&vid=5a15687812137d17cc1e5037082238499bee33c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR