(1S,4S,5S)-4,7,7-Trimethyl-6-thiabicyclo[3.2.1]octane ≥95.0% (by GC)
Supplier: TCI America
T2579-1G
T2579-5G
TCT2579-1GEA
142.2
CAD
TCT2579-1G
TCT2579-5G
(1S,4S,5S)-4,7,7-Trimethyl-6-thiabicyclo[3.2.1]octane ≥95.0% (by GC)
(1S,4S,5S)-4,7,7-Trimethyl-6-thiabicyclo[3.2.1]octane
CAS Number: 1208985-45-8
Molecular Formula: C10H18S
Molecular Weight: 170.31
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific rotation [a]20/D: 67 deg (neat)
Formula:
C₁₀H₁₈S |
CAS Number:
1208985-45-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...