Tetrahydropyran-4-carboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Tetrahydropyran-4-yl-carboxylic acid
T2493-25G
T2493-5G
TCT2493-25GEA
298.62
CAD
TCT2493-25G
TCT2493-5G
Tetrahydropyran-4-carboxylic acid ≥98.0% (by GC, titration analysis)
Tetrahydropyran-4-carboxylic acid
CAS Number: 5337-03-1
MDL Number: MFCD00031016
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 115
Melting point (°C): 89
Formula:
C₆H₁₀O₃ MW: 130.14 g/mol Boiling Pt: 115 °C Melting Pt: 89 °C |
MDL Number:
MFCD00031016 CAS Number: 5337-03-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...