2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl-2,2,2-trichloroacetimidate ≥98.0%

Supplier: TCI America

Synonyms: 2,3,4,6-Tetra-O-acetyl-1-O-trichloroacetimidoyl-beta-D-glucopyranoside, 2,3,4,6-Tetra-O-acetyl-b-D-glucopyranosyl trichloroacetimidate

T2491-1G T2491-5G
TCT2491-1GEA 157.51 CAD
TCT2491-1G TCT2491-5G
2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl-2,2,2-trichloroacetimidate ≥98.0%
2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranosyl-2,2,2-trichloroacetimidate

CAS Number: 92052-29-4
Molecular Formula: C16H20Cl3NO10
Molecular Weight: 492.68
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Melting point (°C): 158
Specific rotation [a]20/D: 7.0 deg (C=1, CHCl3)
Storage Temperature: <0°C

Formula: C₁₆H₂₀Cl₃NO₁₀
Melting Pt: 158 °C
MDL Number: MFCD11112189
CAS Number: 92052-29-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=930430bb7ef4ac778bb69c010883a518265c04a2&vid=930430bb7ef4ac778bb69c010883a518265c04a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR