2-(Trifluoromethoxy)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 2-Trifluoromethoxyphenylboronic acid, 2-(Trifluoromethoxy)benzeneboronic acid, (2-(Trifluoromethoxy)phenyl)metaboric acid, Dihydroxy(2-(trifluoromethoxy)phenyl)borane

T2413-1G T2413-5G
TCT2413-1GEA 28.44 CAD
TCT2413-1G TCT2413-5G
2-(Trifluoromethoxy)phenylboronic acid (contains varying amounts of Anhydride)
2-(Trifluoromethoxy)phenylboronic acid

CAS Number: 175676-65-0
MDL Number: MFCD01320763
Molecular Formula: C7H6BF3O3
Molecular Weight: 205.93
Form: Crystal
Color: White
Melting point (°C): 118


Other Product Names / Synonyms: 2-(Trifluoromethoxy)benzeneboronic Acid

Formula: C₇H₆BF₃O₃
MW: 205.93
Melting Pt: 118 °C
MDL Number: MFCD01320763
CAS Number: 175676-65-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2447244579e2dc0088312ddf7c19cdc2b7a12820&vid=2447244579e2dc0088312ddf7c19cdc2b7a12820&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR