Thiobenzoylmercaptoacetic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
S-(Thiobenzoyl)thioglycolic acid, (Thiobenzoylthio)acetic acid, 2-((Phenylcarbonothioyl)thio)acetic acid
CAS Number: 942-91-6
MDL Number: MFCD00004927
Molecular Formula: C9H8O2S2
Molecular Weight: 212.28
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: Yellow Red
Melting point (°C): 128
Product Categories: Synthetic Organic Chemistry, Thiocarbonyl Compounds, Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: (Thiobenzoylthio)acetic Acid
Formula:
C₉H₈O₂S₂ MW: 212.2886 g/mol Melting Pt: 128 °C |
MDL Number:
MFCD00004927 CAS Number: 942-91-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...