1,2,3-Trifluoro-5-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,2,3-Trifluoro-5-nitrobenzene, 3,4,5-Trifluoronitrobenzene

T2357-25G T2357-5G
TCT2357-25GEA 208.24 CAD
TCT2357-25G TCT2357-5G
1,2,3-Trifluoro-5-nitrobenzene ≥98.0% (by GC)
1,2,3-Trifluoro-5-nitrobenzene

CAS Number: 66684-58-0
MDL Number: MFCD00456803
Molecular Formula: C6H2F3NO2
Molecular Weight: 177.08
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Pale Yellow
Flash Point (°C): 75
Specific Gravity (20/20): 1.54


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,2,3-Trifluoro-5-nitrobenzene

Formula: C₆H₂F₃NO₂
MW: 177.08 g/mol
Density: 1.54 (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00456803
CAS Number: 66684-58-0
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c18a89516ee2be1444c8484119fdf85b4f0a61d4&vid=c269d092c5a5ab7780581accc71d04245d45ccef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR