2,3,4,6-Tetra-O-acetyl-ɑ-D-galactopyranosyl-2,2,2-trichloroacetimidate ≥93.0% (by HPLC)
Supplier: TCI America
T2295-1G
T2295-5G
TCT2295-1GEA
67.94
CAD
TCT2295-1G
TCT2295-5G
2,3,4,6-Tetra-O-acetyl-ɑ-D-galactopyranosyl-2,2,2-trichloroacetimidate ≥93.0% (by HPLC)
2,3,4,6-Tetra-O-acetyl-alpha-D-galactopyranosyl-2,2,2-trichloroacetimidate
CAS Number: 86520-63-0
MDL Number: MFCD07369652
Molecular Formula: C16H20Cl3NO10
Molecular Weight: 492.68
Purity/Analysis Method: >93.0% (HPLC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 122
Specific rotation [a]20/D: 112 deg (C=1, CHCl3)
Storage Temperature: <0°C
Formula:
C₁₆H₂₀Cl₃NO₁₀ Melting Pt: 122 °C |
MDL Number:
MFCD07369652 CAS Number: 86520-63-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...