3,3,5-Trimethylcyclohexyl salicylate (cis and trans mixture) ≥98.0%
Supplier: TCI America
Synonyms:
3,3,5-Trimethylcyclohexyl 2-hydroxybenzoate, Coppertone, 2-Hydroxybenzoic Acid 3,3,5-Trimethylcyclohexyl Ester, Salicylic Acid 3,3,5-Trimethylcyclohexyl Ester, 3,3,5-Trimethylcyclohexyl 2-Hydroxybenzoate, Homosalate
CAS Number: 118-56-9
Molecular Formula: C16H22O3
Molecular Weight: 262.35
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 165
Specific Gravity (20/20): 1.05
Other Product Names / Synonyms: 2-Hydroxybenzoic Acid 3,3,5-Trimethylcyclohexyl Ester, Salicylic Acid 3,3,5-Trimethylcyclohexyl Ester, 3,3,5-Trimethylcyclohexyl 2-Hydroxybenzoate
Formula:
C₁₆H₂₂O₃ MW: 262.35 g/mol Boiling Pt: 165 °C Density: 1.05 (20 °C) |
MDL Number:
MFCD00019377 CAS Number: 118-56-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...