Tetradecyl acrylate ≥95.0% stabilised

Supplier: TCI America

Synonyms: Acrylic Acid Tetradecyl Ester

T2265-25ML T2265-500ML
TCT2265-25MLEA 109.29 CAD
TCT2265-25ML TCT2265-500ML
Tetradecyl acrylate ≥95.0% stabilised
Tetradecyl acrylate

(stabilized with MEHQ)
CAS Number: 21643-42-5
Molecular Formula: C17H32O2
Molecular Weight: 268.44
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 138
Melting point (°C): 14
Specific Gravity (20/20): 0.87


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid Tetradecyl Ester

Formula: C₁₇H₃₂O₂
Boiling Pt: 138 °C
Melting Pt: 14 °C
Density: 0.87 (20 °C)
MDL Number: MFCD00673244
CAS Number: 21643-42-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=088e1435d132e8eb6f82df02a3cbfb4e4b68d09d&vid=088e1435d132e8eb6f82df02a3cbfb4e4b68d09d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR