Triphenylmethyl mercaptan ≥97.0%

Supplier: TCI America

Synonyms: Triphenylmethanethiol

T2244-25G T2244-5G
TCT2244-25GEA 229.1 CAD
TCT2244-25G TCT2244-5G
Triphenylmethyl mercaptan ≥97.0%
Triphenylmethyl mercaptan

CAS Number: 3695-77-0
MDL Number: MFCD00004854
Molecular Formula: C19H16S
Molecular Weight: 276.40
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 108


Other Product Names / Synonyms: Triphenylmethyl Mercaptan, Trityl Mercaptan

Formula: C₁₉H₁₆S
MW: 276.40 g/mol
Melting Pt: 108 °C
MDL Number: MFCD00004854
CAS Number: 3695-77-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59b4ade55356e6d3befaf64a45025feb9fe140f2&vid=59b4ade55356e6d3befaf64a45025feb9fe140f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR