1-Iodo-4-(trifluoromethoxy)benzene ≥98.0% (by GC)

Supplier: TCI America

T2153-25G T2153-5G
TCT2153-25GEA 205.4 CAD
TCT2153-25G TCT2153-5G
1-Iodo-4-(trifluoromethoxy)benzene ≥98.0% (by GC)
1-Iodo-4-(trifluoromethoxy)benzene

CAS Number: 103962-05-6
Molecular Formula: C7H4F3IO
Molecular Weight: 288.01
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Yellow
Boiling point (°C): 97
Flash Point (°C): 78
Specific Gravity (20/20): 1.87

Formula: C₇H₄F₃IO
Boiling Pt: 97 °C
Density: 1.87 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00042411
CAS Number: 103962-05-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af71670fd6cecfb89a6059ebf1cd512b42f0b25e&vid=af71670fd6cecfb89a6059ebf1cd512b42f0b25e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR