Ethyl-2,3,4,5-tetrafluorobenzoate ≥98.0% (by GC)

Supplier: TCI America

T2094-100G T2094-25G
TCT2094-100GEA 171.9 CAD
TCT2094-100G TCT2094-25G
Ethyl-2,3,4,5-tetrafluorobenzoate ≥98.0% (by GC)
Ethyl 2,3,4,5-tetrafluorobenzoate

CAS Number: 122894-73-9
MDL Number: MFCD02094158
Molecular Formula: C9H6F4O2
Molecular Weight: 222.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 88
Specific Gravity (20/20): 1.39


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,3,4,5-Tetrafluorobenzoic Acid Ethyl Ester

Formula: C₉H₆F₄O₂
Boiling Pt: 88 °C
Density: 1.39 (20 °C)
MDL Number: MFCD02094158
CAS Number: 122894-73-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=240c0ac7acdaf4901d4c809bea53a40c426563ca&vid=240c0ac7acdaf4901d4c809bea53a40c426563ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR