1-(Trifluoroacetyl)piperidine ≥98.0% (by GC)
Supplier: TCI America
T2087-25G
T2087-5G
TCT2087-25GEA
175.38
CAD
TCT2087-25G
TCT2087-5G
1-(Trifluoroacetyl)piperidine ≥98.0% (by GC)
1-(Trifluoroacetyl)piperidine
CAS Number: 340-07-8
MDL Number: MFCD00466213
Molecular Formula: C7H10F3NO
Molecular Weight: 181.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 77
Flash Point (°C): 76
Specific Gravity (20/20): 1.24
Formula:
C₇H₁₀F₃NO Boiling Pt: 77 °C Density: 1.24 (20 °C) Flash Pt: 76 °C |
MDL Number:
MFCD00466213 CAS Number: 340-07-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...