2-(Trimethylsilyl)pyridine ≥96.0% (by GC)

Supplier: TCI America

T2021-1G T2021-5G
TCT2021-1GEA 97.96 CAD
TCT2021-1G TCT2021-5G
2-(Trimethylsilyl)pyridine ≥96.0% (by GC)
2-(Trimethylsilyl)pyridine

CAS Number: 13737-04-7
MDL Number: MFCD06797181
Molecular Formula: C8H13NSi
Molecular Weight: 151.28
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 0.92

Formula: C₈H₁₃NSi
Density: 0.92 (20 °C)
MDL Number: MFCD06797181
CAS Number: 13737-04-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36810e560db96aa7af5c92ad87bd48dc2e1ac6e0&vid=36810e560db96aa7af5c92ad87bd48dc2e1ac6e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR