2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
T1943-1G
T1943-5G
TCT1943-1GEA
320.74
CAD
TCT1943-1G
TCT1943-5G
2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid ≥98.0% (by titrimetric analysis)
2,3,5,6-Tetrafluoro-4-hydroxybenzoic acid
CAS Number: 652-34-6
MDL Number: MFCD00129952
Molecular Formula: C7H2F4O3
Molecular Weight: 210.08
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 154
Formula:
C₇H₂F₄O₃ Melting Pt: 154 °C |
MDL Number:
MFCD00129952 CAS Number: 652-34-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...