2,4,6-Trimethylbenzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: (2,4,6-Trimethylphenyl)methanol, Mesitylmethanol

T1905-25G T1905-5G
TCT1905-25GEA 635.16 CAD
TCT1905-25G TCT1905-5G
2,4,6-Trimethylbenzyl alcohol ≥98.0%
2,4,6-Trimethylbenzyl alcohol

CAS Number: 4170-90-5
MDL Number: MFCD00014422
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 141
Melting point (°C): 88


Other Product Names / Synonyms: Mesitylmethanol, 2,4,6-Trimethylbenzenemethanol

Formula: C₁₀H₁₄O
Boiling Pt: 141 °C
Melting Pt: 88 °C
MDL Number: MFCD00014422
CAS Number: 4170-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=412b933f0a22620cac643469faf0f4c1715266b4&vid=412b933f0a22620cac643469faf0f4c1715266b4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR