1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1,4,8,11-Tetramethylcyclam

T1877-1G T1877-200MG
TCT1877-1GEA 293.88 CAD
TCT1877-1G TCT1877-200MG
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane ≥98.0% (by GC, titration analysis)
1,4,8,11-Tetramethyl-1,4,8,11-tetraazacyclotetradecane

CAS Number: 41203-22-9
MDL Number: MFCD00005106
Molecular Formula: C14H32N4
Molecular Weight: 256.44
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 40
Flash Point (°C): 110


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Precise Polymerization Reagents, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 1,4,8,11-Tetramethylcyclam

Formula: C₁₄H₃₂N₄
MW: 256.44 g/mol
Melting Pt: 40 °C
Flash Pt: 110 °C
MDL Number: MFCD00005106
CAS Number: 41203-22-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00cf69834d934c7956041e655a08305fc2f26d91&vid=00cf69834d934c7956041e655a08305fc2f26d91&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR