(2R)-(-)-2,3-Epoxypropyl toluene-4-sulfonate ≥98.0%

Supplier: TCI America

Synonyms: (2R)-(-)-Glycidyl p-toluenesulphonate, (2R)-(-)-Glycidyl tosylate, (2R)-(-)-Glycidyl p-toluenesulfonate

T1611-25G T1611-5G
TCT1611-25GEA 154.84 CAD
TCT1611-25G TCT1611-5G
(2R)-(-)-2,3-Epoxypropyl toluene-4-sulfonate ≥98.0%
(2R)-(-)-2,3-Epoxypropyl toluene-4-sulphonate

CAS Number: 113826-06-5
MDL Number: MFCD00010834
Molecular Formula: C10H12O4S
Molecular Weight: 228.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 48
Flash Point (°C): 202
Specific rotation [a]20/D: -19 deg (C=2.5, CH3CN)


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Chiral Building Blocks
Other Product Names / Synonyms: (2R)-(-)-Glycidyl Tosylate, p-Toluenesulfonic Acid (2R)-(-)-Glycidyl Ester

Formula: C₁₀H₁₂O₄S
MW: 228.2649 g/mol
Melting Pt: 48 °C
Flash Pt: 202 °C
MDL Number: MFCD00010834
CAS Number: 113826-06-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c5c197a79d6b3b39918dd404888bb397b93392b&vid=5c5c197a79d6b3b39918dd404888bb397b93392b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR