Tetrabutylammonium hexafluorophosphate ≥98.0%

Supplier: TCI America

Synonyms: Tetrabutylammonium hexafluorophosphate(V), Tetra-n-butylammonium hexafluorophosphate

T1279-250G T1279-25G
TCT1279-250GEA 684.88 CAD
TCT1279-250G TCT1279-25G
Tetrabutylammonium hexafluorophosphate ≥98.0%
Tetrabutylammonium hexafluorophosphate

CAS Number: 3109-63-5
MDL Number: MFCD00011748
Molecular Formula: C16H36F6NP
Molecular Weight: 387.44
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 245

Formula: C₁₆H₃₆F₆NP
Melting Pt: 245 °C
MDL Number: MFCD00011748
CAS Number: 3109-63-5
UN: 3263
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8957bd1b668522b57fa4f5dee186d43005f301c&vid=c8957bd1b668522b57fa4f5dee186d43005f301c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR