Tris(2-aminoethyl)amine ≥98.0%

Supplier: TCI America

Synonyms: N1,N1-Bis(2-aminoethyl)ethane-1,2-diamine, TREN, 2,2',2''-Triaminotriethylamine

T1243-100ML T1243-5ML
TCT1243-100MLEA 644.64 CAD
TCT1243-100ML TCT1243-5ML
Tris(2-aminoethyl)amine ≥98.0%
Tris(2-aminoethyl)amine

CAS Number: 4097-89-6
MDL Number: MFCD00008177
Molecular Formula: C6H18N4
Molecular Weight: 146.24
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 114
Flash Point (°C): 113
Specific Gravity (20/20): 0.98


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 2,2',2''-Nitrilotriethylamine, 2,2',2''-Triaminotriethylamine, TAEA

Formula: C₆H₁₈N₄
MW: 146.24 g/mol
Boiling Pt: 114 °C
Density: 0.98 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00008177
CAS Number: 4097-89-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8d039233e66c036987ee1d833550e547f523d1f&vid=a8d039233e66c036987ee1d833550e547f523d1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR