Tetrapentylammonium iodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T1011-25G T1011-5G
TCT1011-25GEA 270.18 CAD
TCT1011-25G TCT1011-5G
Tetrapentylammonium iodide ≥98.0% (by titrimetric analysis)
Tetrapentylammonium iodide

CAS Number: 2498-20-6
MDL Number: MFCD00041980
Molecular Formula: C20H44IN
Molecular Weight: 425.48
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 137


Product Categories: Materials Chemistry, Secondary Battery Research Reagents
Other Product Names / Synonyms: Tetrapentylammonium Iodide

Formula: C₂₀H₄₄IN
Melting Pt: 137 °C
MDL Number: MFCD00041980
CAS Number: 2498-20-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=245150758981c440837f15ac69ecbdfea3e4ff24&vid=245150758981c440837f15ac69ecbdfea3e4ff24&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR